3',5'-Dimethyl-2,2,2-trifluoroacetophenone ≥97%
Supplier: AOBChem USA
|
Warning
|
63690-1G
63690-25G
63690-5G
76892-818EA
70.02
USD
76892-818
76892-820
76892-822
3',5'-Dimethyl-2,2,2-trifluoroacetophenone ≥97%
3',5'-Dimethyl-2,2,2-trifluoroacetophenone
Precaución: for Research Use Only
Formula:
C₁₀H₉F₃O Storage Temperature: Ambient |
MDL Number:
MFCD01319988 CAS Number: 132719-10-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...