1-(2,5-Difluorophenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

63453-1G 63453-25G 63453-500MG 63453-5G
76892-802EA 198.39 USD
76892-802 76892-804 76892-806 76892-808
1-(2,5-Difluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(2,5-Difluorophenyl)-2,2,2-trifluoroethanone
Formula: C₈H₃F₅O
MW: 210.1008 g/mol
Storage Temperature: Ambient
CAS Number: 1092712-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR