2,2,2,2',6'-Pentafluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(2,6-Difluorophenyl)-2,2,2-trifluoroethanone, 1-(2,6-Difluorophenyl)-2,2,2-trifluoroethan-1-one

63451-1G 63451-25G 63451-500MG 63451-5G
76892-786EA 284.33 USD
76892-786 76892-788 76892-790 76892-792
2,2,2,2',6'-Pentafluoroacetophenone ≥97%
2,2,2,2',6'-Pentafluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₃F₅O
MW: 210.101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08447307
CAS Number: 1092712-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR