2,4-Dibromo-6-(trifluoromethoxy)aniline ≥97%

Supplier: AOBChem USA

Warning

63436-25G 63436-5G
76892-774EA 204.76 USD
76892-774 76892-776
2,4-Dibromo-6-(trifluoromethoxy)aniline ≥97%
2,4-Dibromo-6-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₄Br₂F₃NO
Storage Temperature: Ambient
CAS Number: 886499-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR