1-(3-(Tert-butyl)phenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

63109-1G 63109-25G 63109-500MG 63109-5G
76892-696EA 211.12 USD
76892-696 76892-698 76892-700 76892-702
1-(3-(Tert-butyl)phenyl)-2,2,2-trifluoroethanone ≥97%
1-(3-(Tert-butyl)phenyl)-2,2,2-trifluoroethanone
Formula: C₁₂H₁₃F₃O
MW: 230.2262 g/mol
Storage Temperature: Ambient
CAS Number: 155628-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR