(5-Chloro-2-fluoro-4-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

63090-1G 63090-25G 63090-500MG 63090-5G
76892-682EA 309.79 USD
76892-682 76892-684 76892-686 76892-688
(5-Chloro-2-fluoro-4-methylphenyl)methanol ≥97%
(5-Chloro-2-fluoro-4-methylphenyl)methanol
Formula: C₈H₈ClFO
MW: 174.5999 g/mol
Storage Temperature: Ambient
CAS Number: 252004-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR