1-Bromo-2-chloro-4-(difluoromethoxy)-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

62964-1G 62964-500MG 62964-5G 62964-25G
76892-660EA 642.91 USD
76892-660 76892-662 76892-664 77656-728
1-Bromo-2-chloro-4-(difluoromethoxy)-3-methylbenzene ≥97%
1-Bromo-2-chloro-4-(difluoromethoxy)-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₆BrClF₂O
MW: 271.4864 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761151
CAS Number: 2168415-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR