1-(3,5-Difluorophenyl)ethanol ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3,5-Difluoro-alpha-methylbenzyl alcohol

62777-25G 62777-5G
76892-634EA 630.18 USD
76892-634 76892-636
1-(3,5-Difluorophenyl)ethanol ≥97% research grade
1-(3,5-Difluorophenyl)ethanol
Formula: C₈H₈F₂O
MW: 158.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038285
CAS Number: 872181-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR