4-Bromo-5-chloro-2-fluoroiodobenzene ≥97%

Supplier: AOBChem USA

Warning

62572-1G 62572-25G 62572-500MG 62572-5G
76892-606EA 183.54 USD
76892-606 76892-608 76892-610 76892-612
4-Bromo-5-chloro-2-fluoroiodobenzene ≥97%
4-Bromo-5-chloro-2-fluoroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClFI
Storage Temperature: Ambient
CAS Number: 1160574-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR