2-Methyl-3-(trifluoromethyl)anisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Methoxy-2-methyl-3-(trifluoromethyl)benzene

62381-1G 62381-25G 62381-5G 62381-500MG
76892-594EA 93.36 USD
76892-594 76892-596 76892-598 77647-830
2-Methyl-3-(trifluoromethyl)anisole ≥97%
2-Methyl-3-(trifluoromethyl)anisole

Precaución: for Research Use Only

Formula: C₉H₉F₃O
MW: 190.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832284
CAS Number: 1017778-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR