(2-(4-(Tert-butoxycarbonyl)piperazin-1-yl)pyridin-4-yl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (2-(4-(tert-Butoxycarbonyl)piperazin-1-yl)pyridin-4-yl)boronic acid

61966-1G 61966-25G 61966-500MG 61966-5G
76892-532EA 206.88 USD
76892-532 76892-534 76892-536 76892-538
(2-(4-(Tert-butoxycarbonyl)piperazin-1-yl)pyridin-4-yl)boronic acid ≥97% research grade
(2-(4-(Tert-butoxycarbonyl)piperazin-1-yl)pyridin-4-yl)boronic acid
Formula: C₁₄H₂₂BN₃O₄
Storage Temperature: Refrigerator
MDL Number: MFCD18261480
CAS Number: 1003043-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR