2-(Benzyloxy)-3,5-dichlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61499-1G 61499-25G 61499-500MG 61499-5G
76892-496EA 250.38 USD
76892-496 76892-498 76892-500 76892-502
2-(Benzyloxy)-3,5-dichlorobenzaldehyde ≥97%
2-(Benzyloxy)-3,5-dichlorobenzaldehyde
Formula: C₁₄H₁₀Cl₂O₂
MW: 281.134 g/mol
Storage Temperature: Ambient
CAS Number: 40359-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR