1-Bromo-3,4,5-trifluoro-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

61062-1G 61062-25G 61062-5G
76892-452EA 66.84 USD
76892-452 76892-454 76892-456
1-Bromo-3,4,5-trifluoro-2-iodobenzene ≥97%
1-Bromo-3,4,5-trifluoro-2-iodobenzene
Formula: C₆HBrF₃I
MW: 336.8758 g/mol
Storage Temperature: Ambient
CAS Number: 202925-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR