1-Bromo-2,5-dichloro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

61034-1G 61034-25G 61034-500MG 61034-5G
76892-436EA 173.99 USD
76892-436 76892-438 76892-440 76892-442
1-Bromo-2,5-dichloro-3-methylbenzene ≥97%
1-Bromo-2,5-dichloro-3-methylbenzene
Formula: C₇H₅BrCl₂
MW: 239.9246 g/mol
Storage Temperature: Ambient
CAS Number: 1000571-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR