2,4-Dibromo-5-(trifluoromethyl)aniline ≥97%

Supplier: AOBChem USA

Warning

60753-1G 60753-25G 60753-500MG 60753-5G
76892-412EA 129.43 USD
76892-412 76892-414 76892-416 76892-418
2,4-Dibromo-5-(trifluoromethyl)aniline ≥97%
2,4-Dibromo-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄Br₂F₃N
Storage Temperature: Ambient
CAS Number: 24115-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR