2,3,6-Trichloro-4-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

60720-25G 60720-5G
76892-400EA 240.83 USD
76892-400 76892-402
2,3,6-Trichloro-4-methylpyridine ≥95%
2,3,6-Trichloro-4-methylpyridine
Formula: C₆H₄Cl₃N
MW: 196.4616 g/mol
Storage Temperature: Ambient
CAS Number: 52137-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR