4'-Chloro-3'-methyl-2,2,2-trifluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

60451-1G 60451-25G 60451-500MG 60451-5G
76892-348EA 185.66 USD
76892-348 76892-350 76892-352 76892-354
4'-Chloro-3'-methyl-2,2,2-trifluoroacetophenone ≥97%
4'-Chloro-3'-methyl-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD02260828
CAS Number: 286017-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR