1-(4-Chloro-2-methoxyphenyl)ethanol ≥97% research grade

Supplier: AOBChem USA

Warning

60383-1G 60383-25G 60383-500MG 60383-5G
76892-332EA 263.11 USD
76892-332 76892-334 76892-336 76892-338
1-(4-Chloro-2-methoxyphenyl)ethanol ≥97% research grade
1-(4-Chloro-2-methoxyphenyl)ethanol
Formula: C₉H₁₁ClO₂
Storage Temperature: Ambient
CAS Number: 1593370-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR