1-Fluoro-3-iodo-2-methoxybenzene ≥97% research grade

Supplier: AOBChem USA

Warning

60236-1G 60236-25G 60236-5G
76892-300EA 63.66 USD
76892-300 76892-302 76892-304
1-Fluoro-3-iodo-2-methoxybenzene ≥97% research grade
1-Fluoro-3-iodo-2-methoxybenzene
Formula: C₇H₆FIO
MW: 252.025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782083
CAS Number: 32750-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR