1-Iodo-2,3-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

60135-1G 60135-25G 60135-5G
76892-260EA 62.6 USD
76892-260 76892-262 76892-264
1-Iodo-2,3-dimethoxybenzene ≥95%
1-Iodo-2,3-dimethoxybenzene
Formula: C₈H₉IO₂
Storage Temperature: Ambient
CAS Number: 25245-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR