3-Fluoro-5-iodobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

59756-1G 59756-25G 59756-500MG 59756-5G
76892-246EA 183.54 USD
76892-246 76892-248 76892-250 76892-252
3-Fluoro-5-iodobenzotrifluoride ≥97%
3-Fluoro-5-iodobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃F₄I
Storage Temperature: Ambient
MDL Number: MFCD09800718
CAS Number: 1027513-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR