(2-Methoxypyrimidin-5-yl)methanol ≥97% research grade

Supplier: AOBChem USA

Warning

59441-1G 59441-25G 59441-500MG 59441-5G
76892-230EA 206.88 USD
76892-230 76892-232 76892-234 76892-236
(2-Methoxypyrimidin-5-yl)methanol ≥97% research grade
(2-Methoxypyrimidin-5-yl)methanol
Formula: C₆H₈N₂O₂
MW: 140.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11840201
CAS Number: 1158735-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR