3-Bromo-2-chloro-6-fluoroiodobenzene ≥97%

Supplier: AOBChem USA

Warning

59320-1G 59320-25G 59320-500MG 59320-5G
76892-214EA 334.19 USD
76892-214 76892-216 76892-218 76892-220
3-Bromo-2-chloro-6-fluoroiodobenzene ≥97%
3-Bromo-2-chloro-6-fluoroiodobenzene
Formula: C₆H₂BrClFI
Storage Temperature: Ambient
MDL Number: MFCD28140548
CAS Number: 1820649-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR