2-(Benzyloxy)-4-chloro-1-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

58977-1G 58977-25G 58977-5G 58977-500MG
76892-192EA 117.76 USD
76892-192 76892-194 76892-196 77651-784
2-(Benzyloxy)-4-chloro-1-iodobenzene ≥97%
2-(Benzyloxy)-4-chloro-1-iodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClIO
MW: 344.575 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27981315
CAS Number: 1820607-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR