1-Bromo-5-chloro-4-fluoro-2-methoxybenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
58293-1G
58293-25G
58293-500MG
58293-5G
76892-132EA
198.39
USD
76892-132
76892-134
76892-136
76892-138
1-Bromo-5-chloro-4-fluoro-2-methoxybenzene ≥97%
1-Bromo-5-chloro-4-fluoro-2-methoxybenzene
Formula:
C₇H₅BrClFO MW: 239.469 g/mol Storage Temperature: Ambient |
CAS Number:
949892-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...