[4-(4-Bromophenoxy)phenyl]methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-(4-Bromophenoxy)phenyl)methanol

57799-1G 57799-25G 57799-500MG 57799-5G
76892-104EA 236.59 USD
76892-104 76892-106 76892-108 76892-110
[4-(4-Bromophenoxy)phenyl]methanol ≥97%
[4-(4-Bromophenoxy)phenyl]methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrO₂
MW: 279.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11190398
CAS Number: 1039899-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR