1-(6-Ethoxypyridin-2-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

57718-1G 57718-25G 57718-500MG 57718-5G
76892-092EA 436.03 USD
76892-092 76892-094 76892-096 76892-098
1-(6-Ethoxypyridin-2-yl)ethanone ≥97%
Formula: C₉H₁₁NO₂
Storage Temperature: Ambient
CAS Number: 21190-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR