2,4-Dibromo-6-fluorobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

57697-1G 57697-25G 57697-500MG 57697-5G
76892-076EA 206.88 USD
76892-076 76892-078 76892-080 76892-082
2,4-Dibromo-6-fluorobenzonitrile ≥97%
2,4-Dibromo-6-fluorobenzonitrile
Formula: C₇H₂Br₂FN
Storage Temperature: Ambient
MDL Number: MFCD22377850
CAS Number: 1393585-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR