1,4-Dichloro-2-(methylsulfonyl)benzene ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2,5-Dichlorophenyl methyl sulphone

57688-1G 57688-25G 57688-5G 57688-500MG
76892-062EA 96.55 USD
76892-062 76892-064 76892-066 77651-740
1,4-Dichloro-2-(methylsulfonyl)benzene ≥97% research grade
1,4-Dichloro-2-(methylsulfonyl)benzene
Formula: C₇H₆Cl₂O₂S
MW: 225.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03789183
CAS Number: 66640-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR