(2-Fluoro-4-methoxypyridin-3-yl)methanol ≥97% research grade

Supplier: AOBChem USA

Warning

57364-1G 57364-25G 57364-500MG 57364-5G
76892-036EA 355.41 USD
76892-036 76892-038 76892-040 76892-042
(2-Fluoro-4-methoxypyridin-3-yl)methanol ≥97% research grade
(2-Fluoro-4-methoxypyridin-3-yl)methanol
Formula: C₇H₈FNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD18819101
CAS Number: 451459-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR