1-Isobutyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

57260-1G 57260-25G 57260-500MG 57260-5G
76892-022EA 245.07 USD
76892-022 76892-024 76892-026 76892-028
1-Isobutyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde ≥97%
1-Isobutyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde
Formula: C₁₀H₁₆N₂O
MW: 180.247 g/mol
Storage Temperature: Refrigerator
CAS Number: 1020519-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR