1-Isobutyl-3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

57257-1G 57257-500MG 57257-5G
76892-008EA 553.79 USD
76892-008 76892-010 76892-012
1-Isobutyl-3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥97%
1-Isobutyl-3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formula: C₁₅H₂₇BN₂O₂
MW: 278.198 g/mol
Storage Temperature: Ambient
CAS Number: 2205068-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR