1-Isopropyl-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

57249-1G 57249-25G 57249-5G 57249-500MG
76891-996EA 117.76 USD
76891-996 76891-998 76892-000 77651-730
1-Isopropyl-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥97%
1-Isopropyl-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
Formula: C₉H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06260701
CAS Number: 1007542-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR