2-Chloro-5-iodoanisole ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-Chloro-4-iodo-2-methoxybenzene

56781-1G 56781-25G 56781-500MG 56781-5G
76891-944EA 198.39 USD
76891-944 76891-946 76891-948 76891-950
2-Chloro-5-iodoanisole ≥95%
2-Chloro-5-iodoanisole
Formula: C₇H₆ClIO
MW: 268.47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00672968
CAS Number: 161949-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR