2,4-Difluoro-5-iodoanisole ≥97%

Supplier: AOBChem USA

Warning

56648-1G 56648-25G 56648-500MG 56648-5G
76891-930EA 206.88 USD
76891-930 76891-932 76891-934 76891-936
2,4-Difluoro-5-iodoanisole ≥97%
2,4-Difluoro-5-iodoanisole
Formula: C₇H₅F₂IO
Storage Temperature: Ambient
MDL Number: MFCD28986200
CAS Number: 1936250-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR