1-Phenyl-1H-pyrazole-5-carbaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
56563-1G
56563-25G
56563-5G
76891-916EA
79.57
USD
76891-916
76891-918
76891-920
1-Phenyl-1H-pyrazole-5-carbaldehyde ≥97%
1-Phenyl-1H-pyrazole-5-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₀H₈N₂O MW: 172.1800 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00103497 CAS Number: 132274-70-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...