(2-Bromo-6-fluoro-5-iodo-3-methoxyphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

56043-1G 56043-25G 56043-500MG 56043-5G
76891-852EA 266.29 USD
76891-852 76891-854 76891-856 76891-858
(2-Bromo-6-fluoro-5-iodo-3-methoxyphenyl)trimethylsilane ≥97%
(2-Bromo-6-fluoro-5-iodo-3-methoxyphenyl)trimethylsilane
Formula: C₁₀H₁₃BrFIOSi
MW: 403.102 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34564339
CAS Number: 2734772-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR