1-Methyl-5-(methylthio)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

55976-25G 55976-5G
76891-840EA 240.83 USD
76891-840 76891-842
1-Methyl-5-(methylthio)-1H-pyrazole ≥97%
1-Methyl-5-(methylthio)-1H-pyrazole
Formula: C₅H₈N₂S
MW: 128.195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530469
CAS Number: 87110-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR