1,3-Difluoro-5-iodo-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

55488-1G 55488-25G 55488-500MG 55488-5G
76891-794EA 233.4 USD
76891-794 76891-796 76891-798 76891-800
1,3-Difluoro-5-iodo-2-methylbenzene ≥97%
1,3-Difluoro-5-iodo-2-methylbenzene
Formula: C₇H₅F₂I
MW: 254.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922635
CAS Number: 1208074-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR