1-(4-(Tert-butyl)phenyl)-2,2-dichloroethanol ≥97%

Supplier: AOBChem USA

Warning

55130-1G 55130-25G 55130-500MG 55130-5G
76891-768EA 227.04 USD
76891-768 76891-770 76891-772 76891-774
1-(4-(Tert-butyl)phenyl)-2,2-dichloroethanol ≥97%
1-(4-(Tert-butyl)phenyl)-2,2-dichloroethanol
Formula: C₁₂H₁₆Cl₂O
MW: 247.161 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34564287
CAS Number: 59477-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR