1-Bromo-2-chloro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

54958-1G 54958-25G 54958-500MG 54958-5G
76891-736EA 242.95 USD
76891-736 76891-738 76891-740 76891-742
1-Bromo-2-chloro-3-iodobenzene ≥97%
1-Bromo-2-chloro-3-iodobenzene
Formula: C₆H₃BrClI
Storage Temperature: Ambient
MDL Number: MFCD18379733
CAS Number: 57012-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR