2-Bromo-4-fluoro-5-iodotoluene ≥97%

Supplier: AOBChem USA

Warning

54908-1G 54908-25G 54908-5G
76891-706EA 79.57 USD
76891-706 76891-708 76891-710
2-Bromo-4-fluoro-5-iodotoluene ≥97%
2-Bromo-4-fluoro-5-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrFI
Storage Temperature: Ambient
MDL Number: MFCD09800683
CAS Number: 677777-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR