2-Bromo-5-chloro-4-fluorotoluene ≥97%

Supplier: AOBChem USA

Warning

54868-1G 54868-25G 54868-5G 54868-500MG
76891-692EA 109.28 USD
76891-692 76891-694 76891-696 77651-616
2-Bromo-5-chloro-4-fluorotoluene ≥97%
2-Bromo-5-chloro-4-fluorotoluene
Formula: C₇H₅BrClF
Storage Temperature: Refrigerator
MDL Number: MFCD20441895
CAS Number: 1067882-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR