1-Bromo-3-ethyl-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

54823-1G 54823-25G 54823-500MG 54823-5G
76891-676EA 369.2 USD
76891-676 76891-678 76891-680 76891-682
1-Bromo-3-ethyl-5-methylbenzene ≥97%
1-Bromo-3-ethyl-5-methylbenzene
Formula: C₉H₁₁Br
MW: 199.088 g/mol
Storage Temperature: Ambient
CAS Number: 1208075-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR