2,2-Dichloro-1-(4-fluorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

54530-1G 54530-25G 54530-500MG 54530-5G
76891-608EA 228.1 USD
76891-608 76891-610 76891-612 76891-614
2,2-Dichloro-1-(4-fluorophenyl)ethanol ≥97%
2,2-Dichloro-1-(4-fluorophenyl)ethanol
Formula: C₈H₇Cl₂FO
Storage Temperature: Refrigerator
MDL Number: MFCD22044381
CAS Number: 61693-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR