1-Bromo-4-iodo-2-methoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

54442-1G 54442-25G 54442-500MG 54442-5G
76891-594EA 216.43 USD
76891-594 76891-596 76891-598 76891-600
1-Bromo-4-iodo-2-methoxy-5-methylbenzene ≥97%
1-Bromo-4-iodo-2-methoxy-5-methylbenzene
Formula: C₈H₈BrIO
Storage Temperature: Ambient
CAS Number: 1887767-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR