2'-Methoxy-5'-(trifluoromethyl)acetophenone ≥95%
Supplier: AOBChem USA
|
Warning
|
54241-1G
54241-25G
54241-5G
76891-568EA
70.02
USD
76891-568
76891-570
76891-572
2'-Methoxy-5'-(trifluoromethyl)acetophenone ≥95%
2'-Methoxy-5'-(trifluoromethyl)acetophenone
Precaución: for Research Use Only
Formula:
C₁₀H₉F₃O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD09832300 CAS Number: 503464-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...