1-(4-Bromo-1H-pyrazol-1-yl)-2-methylpropan-2-ol ≥97%

Supplier: AOBChem USA

Warning

53944-1G 53944-500MG 53944-5G 53944-25G
76891-538EA 492.26 USD
76891-538 76891-540 76891-542 77656-462
1-(4-Bromo-1H-pyrazol-1-yl)-2-methylpropan-2-ol ≥97%
1-(4-Bromo-1H-pyrazol-1-yl)-2-methylpropan-2-ol
Formula: C₇H₁₁BrN₂O
MW: 219.079 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12779230
CAS Number: 1008510-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR