1,5-Dibromo-2-chloro-4-fluorobenzene 97%

Supplier: AOBChem USA

Warning

53873-1G 53873-25G 53873-5G 53873-500MG
76891-526EA 88.06 USD
76891-526 76891-528 76891-530 77651-568
1,5-Dibromo-2-chloro-4-fluorobenzene 97%
1,5-Dibromo-2-chloro-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂ClF
Storage Temperature: Ambient
MDL Number: MFCD09878127
CAS Number: 1000578-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR