1,2-Difluoro-4-iodo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

53826-1G 53826-25G 53826-500MG 53826-5G
76891-512EA 292.81 USD
76891-512 76891-514 76891-516 76891-518
1,2-Difluoro-4-iodo-3-methylbenzene ≥97%
1,2-Difluoro-4-iodo-3-methylbenzene
Formula: C₇H₅F₂I
Storage Temperature: Ambient
MDL Number: MFCD12922591
CAS Number: 1208078-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR